(R)-(+)-1-Amino-2-(methoxymethyl)pyrrolidine 95+%

Supplier: Thermo Scientific Chemicals
Warning

27714-0010
TS27714-0010EA 167 USD
TS27714-0010
(R)-(+)-1-Amino-2-(methoxymethyl)pyrrolidine 95+%
(R)-(+)-1-Amino-2-(methoxymethyl)pyrrolidine
Formula: C₆H₁₄N₂O
Boiling Pt: 42 °C (1.8 mmHg)
Density: 0.97 g/cm³
Flash Pt: 72 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010622
CAS Number: 72748-99-3

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4635 to 1.4655 (20°C, 589 nm)
GC ≥95.0 %
Specific optical rotation +76° to +84° (20°C, 589 nm) (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR