2-Methylindoline 99+%

Supplier: Thermo Scientific Chemicals
Warning

27588-0050 27588-1000
TS27588-0050EA 31 USD
TS27588-0050 TS27588-1000
2-Methylindoline 99+%
2-Methylindoline
Formula: C₉H₁₁N
MW: 133.19 g/mol
Boiling Pt: 228
Melting Pt: –51 °C
Density: 1.02 g/cm³
Flash Pt: 103 °C
MDL Number: MFCD00005706
CAS Number: 6872-06-6
EINECS: 229-971-0

Order Now

Specification Test Results

Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5680 to 1.5700 (20°C, 589 nm)
Water ≤0.5 % (K.F.)
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR