N-(tert-Butoxycarbonyl)-L-phenylalanine 99+%

Supplier: Thermo Scientific Chemicals

27564-0050 27564-0250 27564-1000
TS27564-0050EA 30 USD
TS27564-0050 TS27564-0250 TS27564-1000
N-(tert-Butoxycarbonyl)-L-phenylalanine 99+%
N-(tert-Butoxycarbonyl)-L-phenylalanine
Formula: C₁₄H₁₉NO₄
Melting Pt: 85…88 °C
Storage Temperature: Ambient
MDL Number: MFCD00002663
CAS Number: 13734-34-4

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 85 to 88 °C
Water ≤0.5 % (K.F.)
HPLC ≥99.0 %
Specific optical rotation +24° to +26° (20 °C, 589 nm) (c=1, ethanol)
Titration with NaOH ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR