β-(N-Morpholino)ethanesulfonic acid (MES), (max. 1% H₂O) 99+%

Supplier: Thermo Scientific Chemicals

27414-0010 27414-0500 27414-2500
TS27414-0010EA 1001 USD
TS27414-0010 TS27414-0500 TS27414-2500
β-(N-Morpholino)ethanesulfonic acid (MES), (max. 1% H₂O) 99+%
β-(N-Morpholino)ethanesulphonic acid (MES)
Formula: C₆H₁₃NO₄S
Melting Pt: >300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006181
CAS Number: 4432-31-9

Order Now

Specification Test Results

Appearance White Crystals
Infrared spectrum Conforms
Water ≤1.0 % (K.F.)
Solubility (0.5 M in water) Clear colorless
Titration with NaOH ≥99.0 % (On anhydrous basis)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR