1,2:5,6-Di-O-isopropylidene-α-D-allofuranose 98%

Supplier: Thermo Scientific Chemicals

27096-0100
TS27096-0100EA 341 USD
TS27096-0100
1,2:5,6-Di-O-isopropylidene-α-D-allofuranose 98%
1,2:5,6-Di-O-isopropylidene-alpha-D-allofuranose
Formula: C₁₂H₂₀O₆
Melting Pt: 73…77 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00135634
CAS Number: 2595-05-3

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 72 to 77 °C
GC ≥97.5 %
Specific optical rotation +33.0° to +39.0° (20 °C, 589 nm) (c=1, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR