Nickel(II) chloride hexahydrate 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Nickel dichloride hexahydrate

AC27051-0010 AC27051-0050 AC27051-2500
TS27051-0010EA 264 USD
TS27051-0010 TS27051-0050 TS27051-2500
Nickel(II) chloride hexahydrate 98+%
Nickel (II) chloride hexahydrate
Formula: NiCl₂·6H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149809
CAS Number: 7791-20-0
EINECS: 231-743-0
UN: 3288
ADR: 6.1,III
Merck Index: 13,06529

Order Now

Specification Test Results

Appearance Green Crystalline powder or crystals
Titration Complexometric ≥98.0 %
pH 4 to 6 (5 % at 20°C)
Sulfate (SO4) ≤50 ppm
Cadmium (Cd) ≤10 ppm
Calcium (Ca) ≤50 ppm
Cobalt (Co) ≤60 ppm
Copper (Cu) ≤10 ppm
Iron (Fe) ≤20 ppm
Lead (Pb) ≤10 ppm
Magnesium (Mg) ≤10 ppm
Sodium (Na) ≤50 ppm
Zinc (Zn) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR