Potassium chloride 99.999% (trace metals basis)

Supplier: Thermo Scientific Chemicals

AC26896-0250 AC26896-1000
TS26896-0250EA 97 USD
TS26896-0250 TS26896-1000
Potassium chloride 99.999% (trace metals basis)
Potassium chloride
Formula: KCl
Boiling Pt: 1420 °C (760 mmHg)
Melting Pt: 770 °C
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Total trace metal impurities ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR