Thallium(I) nitrate 99.5%
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC26559-0250
AC26559-1000
TS26559-0250EA
94
USD
TS26559-0250
TS26559-1000
Thallium(I) nitrate 99.5%
Thallium (I) nitrate
Formula:
NO₃Tl Boiling Pt: 433 °C (760 mmHg) Melting Pt: 206 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011280 CAS Number: 10102-45-1 EINECS: 233-273-1 UN: 2727 ADR: 6.1,II Merck Index: 12,09400 |
Specification Test Results
Appearance | White Crystalline powder or crystals |
Total trace metal impurities | ≤5000 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...