Thallium(I) nitrate 99.5%

Supplier: Thermo Scientific Chemicals
Danger

AC26559-0250 AC26559-1000
TS26559-0250EA 94 USD
TS26559-0250 TS26559-1000
Thallium(I) nitrate 99.5%
Thallium (I) nitrate
Formula: NO₃Tl
Boiling Pt: 433 °C (760 mmHg)
Melting Pt: 206 °C
Storage Temperature: Ambient
MDL Number: MFCD00011280
CAS Number: 10102-45-1
EINECS: 233-273-1
UN: 2727
ADR: 6.1,II
Merck Index: 12,09400

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Total trace metal impurities ≤5000 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR