Sodium hydroxide 50% (w/w) in water, extra pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye
259860010
259860025
259860050
259860250
259860251
TS259860010EA
67
USD
TS259860010
TS259860025
TS259860050
TS259860250
TS259860251
Sodium hydroxide 50% (w/w) in water, extra pure
Sodium hydroxide
Formula:
NaOH Boiling Pt: 119…135 °C (1013 hPa) Melting Pt: 12 °C Density: 1.5 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1824 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless Viscous solution |
Heavy metals | ≤10 ppm |
Density | (20°C) 1.510 to 1.540 kg/l |
Titration with HCl | 49.0 to 51.0 % |
Sodium carbonate (Na2CO3) | ≤0.2 % (Na2CO3) |
Sodium sulfate (Na2SO4) | ≤100 ppm (Na2SO4) |
Sodium chloride (NaCl) | ≤200 ppm (NaCl) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...