Acetonitrile 99.5% for analysis

Supplier: Thermo Scientific Chemicals
Danger

25856-0025 258560010 258560250
TS25856-0025EA 201 USD
TS25856-0025 TS258560010 TS258560250
Acetonitrile 99.5% for analysis
Acetonitrile
Formula: H₃CCN
Boiling Pt: 81…82 °C (760 mmHg)
Melting Pt: –46 °C
Density: 0.781 g/cm³
Flash Pt: 2 °C
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.1 % (Coulometric)
GC ≥99.4 %
Color scale ≤20 APHA
Residue after evaporation ≤0.005 %
Calcium (Ca) ≤1 ppm
Copper (Cu) ≤2 ppm
Iron (Fe) ≤2 ppm
Lead (Pb) ≤2 ppm
Magnesium (Mg) ≤0.5 ppm
Potassium (K) ≤1 ppm
Sodium (Na) ≤5 ppm
Zinc (Zn) ≤2 ppm
Free acid (CH3COOH) ≤0.005 %
Free alkali (NH3) ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR