Methyl cellulose, 4000 mPa.S (2% H₂O)

Supplier: Thermo Scientific Chemicals

Synonyms: Cellulose methyl ether

25811-0025 25811-1000 25811-5000
TS25811-0025EA 423 USD
TS25811-0025 TS25811-1000 TS25811-5000
Methyl cellulose, 4000 mPa.S (2% H₂O)
Methyl cellulose
Storage Temperature: Ambient
MDL Number: MFCD00081763
CAS Number: 9004-67-5

Order Now

Specification Test Results

Appearance White to yellowish-white to greyish-white Powder
Infrared spectrum Conforms
Loss on drying ≤5 %
Viscosity 3000 to 5600 cP (2 % in water at 20°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR