Mercury(II) acetate 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Mercury diacetate, Mercuric acetate, Acetic acid mercury (II) salt

AC25565-0010 AC25565-0050 AC25565-1000
TS25565-0010EA 973 USD
TS25565-0010 TS25565-0050 TS25565-1000
Mercury(II) acetate 99+%
Mercury (II) acetate
Formula: (H₃CCOO)₂Hg
Melting Pt: 179…182 °C
Storage Temperature: Ambient
MDL Number: MFCD00012165
CAS Number: 1600-27-7
EINECS: 216-491-1
UN: 1629
ADR: 6.1,II
Merck Index: 13,05898

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Titration Complexometric ≥99.0 %
Residue after ignition ≤0.02 % (substances not reducible with formic acid)
Chloride (Cl) ≤50 ppm
Nitrate (NO3) ≤50 ppm
Sulfate (SO4) ≤50 ppm
Iron (Fe) ≤10 ppm
Heavy metals (as Pb) ≤20 ppm
Mercurous Mercury (as Hg) ≤4000 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR