Ethoxyquin

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Ethoxy-1,2-dihydro-2,2,4-trimethylquinoline, 6-Ethoxy-2,2,4-trimethyl-1,2-dihydroquinoline, 1,2-Dihydro-6-ethoxy-2,2,4-trimethylquinoline, Ethoxyquin

25563-0250
TS25563-0250EA 30 USD
TS25563-0250 TS25563-1000
Ethoxyquin
Ethoxyquin
Formula: C₁₄H₁₉NO
Boiling Pt: 161 °C (1.6kPa)
Flash Pt: 137 °C
Storage Temperature: Ambient
MDL Number: MFCD00023883
CAS Number: 91-53-2

Order Now

Specification Test Results

Appearance Yelow to red to black Liquid
Infrared spectrum Conforms
Refractive index 1.5720 to 1.5750 (20°C, 589 nm)
GC ≥88 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR