L(+)-Calcium lactate pentahydrate 98%

Supplier: Thermo Scientific Chemicals

25020-0010
TS25020-0010EA 239 USD
TS25020-0010
L(+)-Calcium lactate pentahydrate 98%
L(+)-Calcium lactate pentahydrate
Formula: C₆H₁₀CaO₆·5H₂O
Melting Pt: 240 °C
Storage Temperature: Ambient
MDL Number: MFCD00150344
CAS Number: 5743-47-5
EINECS: 248-953-3

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Titration Complexometric ≥97.5 % (on dry substance)
Loss on drying 22.0 % to 27.0 %
Heavy metals ≤10 ppm
pH 6.0 to 8.0 (5 % in water)
Chloride (Cl) ≤80 ppm
Phosphate (PO4) ≤150 ppm
Sulfate (SO4) ≤400 ppm
Arsenic (As) ≤1 ppm
Calcium (Ca) 13.4 to 14.5 %
Iron (Fe) ≤40 ppm
Alkalies+Mg (as sulfates) ≤1.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR