Ethyl-2-(bromomethyl)acrylate 97%

Supplier: Thermo Scientific Chemicals
Danger

23536-0010 23536-0050
TS23536-0010EA 66 USD
TS23536-0010 TS23536-0050
Ethyl-2-(bromomethyl)acrylate 97%
Ethyl-2-(bromomethyl)acrylate

Appearance: Clear colorless to light yellow Liquid

Formula: C₆H₉BrO₂
Boiling Pt: 86 °C (20 mmHg)
Density: 1.38 g/cm³
Flash Pt: 81 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00031518
CAS Number: 17435-72-2
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4770 to 1.4810 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR