Ethyl-2-(bromomethyl)acrylate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
23536-0010
23536-0050
TS23536-0010EA
66
USD
TS23536-0010
TS23536-0050
Ethyl-2-(bromomethyl)acrylate 97%
Ethyl-2-(bromomethyl)acrylate
Appearance: Clear colorless to light yellow Liquid
Formula:
C₆H₉BrO₂ Boiling Pt: 86 °C (20 mmHg) Density: 1.38 g/cm³ Flash Pt: 81 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00031518 CAS Number: 17435-72-2 UN: 3265 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4770 to 1.4810 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...