Prostaglandin E2 98%

Supplier: Thermo Scientific Chemicals
Warning

23356-0100
TS23356-0100EA 588 USD
TS23356-0100
Prostaglandin E2 98%
Prostaglandin E2
Formula: C₂₀H₃₂O₅
Melting Pt: 66…68 °C
Storage Temperature: Freezer
MDL Number: MFCD00077861
CAS Number: 363-24-6

Order Now

Specification Test Results

Appearance White to cream to light yellow Powder
Infrared spectrum Conforms
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR