Iron(II) sulfide, fused sticks

Supplier: Thermo Scientific Chemicals

Synonyms: Ferrous sulphide, Ferrous sulfide

AC23330-0010
TS23330-0010EA 62 USD
TS23330-0010
Iron(II) sulfide, fused sticks
Iron (II) sulphide
Formula: FeS
Melting Pt: 1195 °C
Density: 4.84 g/cm³
MDL Number: MFCD00011013
CAS Number: 1317-37-9
EINECS: 215-268-6
Merck Index: 12,04106

Order Now

Specification Test Results

Appearance Gray-black Sticks
Assay (S) about 25%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR