Sodium dodecyl sulfate (SDS) 99% for biochemistry

Supplier: Thermo Scientific Chemicals
Danger

23042-0025 23042-1000 23042-5000 23042-0100 23042-0250
TS23042-0025EA 871 USD
TS23042-0025 TS23042-1000 TS23042-5000 TS23042-0100 TS23042-0250
Sodium dodecyl sulfate (SDS) 99% for biochemistry
Sodium dodecyl sulphate (SDS)
Formula: CH₃(CH₂)₁₁OSO₃Na
Melting Pt: 206 °C
Flash Pt: >150 °C
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
UN: 1325
ADR: 4.1,III
Merck Index: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder or flakes
Infrared spectrum Conforms
Loss on drying ≤1 % (1 g, 105°C, 3 h)
Solubility (3 % in water) Clear colorless
GC ≥98.5 %
UV at 280 nm A: ≤0.04
Chloride (Cl) ≤500 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR