Sodium dodecyl sulfate (SDS) 99% for biochemistry
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Sodium lauryl sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, Dodecyl sulfuric acid sodium salt, Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate
23042-0025
23042-1000
23042-5000
23042-0100
23042-0250
TS23042-0025EA
871
USD
TS23042-0025
TS23042-1000
TS23042-5000
TS23042-0100
TS23042-0250
Sodium dodecyl sulfate (SDS) 99% for biochemistry
Sodium dodecyl sulphate (SDS)
Formula:
CH₃(CH₂)₁₁OSO₃Na Melting Pt: 206 °C Flash Pt: >150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00036175 CAS Number: 151-21-3 EINECS: 205-788-1 UN: 1325 ADR: 4.1,III Merck Index: 14,08636 |
|
Specification Test Results
Appearance | White Crystalline powder or flakes |
Infrared spectrum | Conforms |
Loss on drying | ≤1 % (1 g, 105°C, 3 h) |
Solubility | (3 % in water) Clear colorless |
GC | ≥98.5 % |
UV | at 280 nm A: ≤0.04 |
Chloride (Cl) | ≤500 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...