Sodium dodecyl sulfate (SDS) 99% for biochemistry

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium lauryl sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, Dodecyl sulfuric acid sodium salt, Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate

23042-0025 23042-1000 23042-5000 23042-0100 23042-0250
TS23042-0025EA 871 USD
TS23042-0025 TS23042-1000 TS23042-5000 TS23042-0100 TS23042-0250
Sodium dodecyl sulfate (SDS) 99% for biochemistry
Sodium dodecyl sulphate (SDS)
Formula: CH₃(CH₂)₁₁OSO₃Na
Melting Pt: 206 °C
Flash Pt: >150 °C
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
UN: 1325
ADR: 4.1,III
Merck Index: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder or flakes
Infrared spectrum Conforms
Loss on drying ≤1 % (1 g, 105°C, 3 h)
Solubility (3 % in water) Clear colorless
GC ≥98.5 %
UV at 280 nm A: ≤0.04
Chloride (Cl) ≤500 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR