N-Octyl-β-D-glucopyranoside 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Octylglucopyranoside, OG, (2R,3S,4S,5R,6R)-2-(Hydroxymethyl)-6-(octyloxy)tetrahydro-2H-pyran-3,4,5-triol, n-Octyl β-D-glucopyranoside, 1-O-n-Octyl-β-D-glucopyranoside, Octyl-β-D-glucopyranoside

23034-0010 23034-0050 23034-0250
TS23034-0010EA 133 USD
TS23034-0010 TS23034-0050 TS23034-0250 TS23034-2500
N-Octyl-β-D-glucopyranoside 98%
N-Octyl-β-D-glucopyranoside
Formula: C₁₄H₂₈O₆
MW: 292.37 g/mol
Melting Pt: 110
Storage Temperature: Freezer
MDL Number: MFCD00063288
CAS Number: 29836-26-8
EINECS: 249-887-8

Order Now

Specification Test Results

Appearance White to off-white Powder and/or chunks
Infrared spectrum Conforms
Solubility (10 % in water) Clear to slightly hazy colorless to light yellow
HPLC ≥97.5 % (ELSD)
Specific optical rotation -31° to -35° (20°C, 589 nm) (c=5, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR