Dexamethasone 96%

Supplier: Thermo Scientific Chemicals
Warning

23030-0010 23030-0050 23030-2500
TS23030-0010EA 329 USD
TS23030-0010 TS23030-0050 TS23030-2500
Dexamethasone 96%
Dexamethasone
Formula: C₂₂H₂₉FO₅
Melting Pt: 255…261 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064136
CAS Number: 50-02-2
EINECS: 200-003-9

Order Now

Specification Test Results

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (500 mg, 105°C)
UV spectrophotometry ≥95 % (On dried substance)
Related substances (HPLC) Passes test
Specific optical rotation +86° to +92° (20°C, 589 nm) (c=1, ethanol) (On dried substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR