D-(+)-Biotin 98%

Supplier: Thermo Scientific Chemicals

23009-0250 23009-0010 23009-0050 23009-5000
TS23009-0250EA 1330 USD
TS23009-0250 TS23009-0010 TS23009-0050 TS23009-5000
D-(+)-Biotin 98%
D(+)-Biotin
Formula: C₁₀H₁₆N₂O₃S
Melting Pt: 231…233 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00005541
CAS Number: 58-85-5
EINECS: 200-399-3
Merck Index: 13,01231

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤1 % (105°C)
Sulfated ash ≤0.1 %
Appearance of solution (2% soln. in 0.1 N NaOH) clear, colorless
Specific optical rotation +89° to +93° (20°C, 589 nm) (c=2, 0.1 N NaOH) (on dried substance)
Titration with NaOH ≥97.5 % (On dried substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR