Orange G 85+%, pure, high purity biological stain

Supplier: Thermo Scientific Chemicals

Synonyms: Disodium 7-hydroxy-8-phenylazonaphthalene-1,3-disulphonate, Wool Orange 2G, Orange G sodium salt, CI 16230, Acid Orange 10, Disodium 7-hydroxy-8-phenylazonaphthalene-1,3-disulfonate

22982-0250 22982-1000
TS22982-0250EA 55 USD
TS22982-0250 TS22982-1000
Orange G 85+%, pure, high purity biological stain
Orange G
Formula: C₁₆H₁₀N₂Na₂O₇S₂
MW: 452.38 g/mol
Melting Pt: 141
Storage Temperature: Ambient
MDL Number: MFCD00012457
CAS Number: 1936-15-8
EINECS: 217-705-6

Order Now

Specification Test Results

Appearance Orange to orange-red Powder
Infrared spectrum Conforms
UV spectrum Complies
Dye content ≥85.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR