Neutral red, pure, high purity biological stain

Supplier: Thermo Scientific Chemicals

22981-0250 22981-1000
TS22981-0250EA 84 USD
TS22981-0250 TS22981-1000
Neutral red, pure, high purity biological stain
Neutral red
Formula: C₁₅H₁₇ClN₄
Melting Pt: 290 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012651
CAS Number: 553-24-2
EINECS: 209-035-8
Merck Index: 1206571

Order Now

Specification Test Results

Appearance Dark green Crystalline powder
Infrared spectrum Conforms
Dye content ≥50 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR