D-Cycloserine 98%

Supplier: Thermo Scientific Chemicals

22848-0050 22848-0250 22848-0010
TS22848-0050EA 293 USD
TS22848-0050 TS22848-0250 TS22848-0010
D-Cycloserine 98%
D-Cycloserine
Formula: C₃H₆N₂O₂
Melting Pt: 147 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00005353
CAS Number: 68-41-7
EINECS: 200-688-4

Order Now

Specification Test Results

Appearance White to light yellow Powder or crystals
Infrared spectrum Conforms
Loss on drying ≤1 % (60°C, 3 hrs) (vacuum)
UV at 285 nm A: ≤0.8
Condensation products (0.04 % in 0.1 N NaOH)
Potency ≥900 µg/mg
Residue after ignition ≤0.5 %
pH 5.5 to 6.5 (10 % solution)
Specific optical rotation +108° to +114° (20°C, 589 nm) (c=5, 2N NaOH)
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR