Cetrimonium bromide 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N-Cetyl-N,N,N-trimethylammonium bromide, Cetyltrimethylammonium bromide, Hexadecyltrimethylammonium bromide, CTAB, (1-Hexadecyl)trimethylammonium bromide, N,N,N-Trimethylhexadecan-1-aminium bromide

22716-0025 22716-5000 22716-0100 22716-0101 22716-0250 22716-1000
TS22716-0025EA 823 USD
TS22716-0025 TS22716-5000 TS22716-0100 TS22716-0101 TS22716-0250 TS22716-1000
Cetrimonium bromide 99+%
Cetrimonium bromide
Formula: CH₃(CH₂)₁₅N(Br)(CH₃)₃
Melting Pt: 230 °C
Flash Pt: 244 °C
Storage Temperature: Ambient
MDL Number: MFCD00011772
CAS Number: 57-09-0
EINECS: 200-311-3
UN: 3077
ADR: 9,III
Merck Index: 13,02034

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR