D-(-)-Serine 98%

Supplier: Thermo Scientific Chemicals

Synonyms: (R)-(-)-Serine, H-D-Ser-OH

22707-0050 22707-0250 22707-1000
TS22707-0050EA 52 USD
TS22707-0050 TS22707-0250 TS22707-1000
D-(-)-Serine 98%
D(-)-Serine
Formula: C₃H₇NO₃
Melting Pt: 220 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00004269
CAS Number: 312-84-5
EINECS: 206-229-4
Merck Index: 12,08604

Order Now

Specification Test Results

Appearance White to off-white Powder or crystals
Infrared spectrum Conforms
Loss on drying ≤0.3 % (105°C, 3 h)
Sulfated ash ≤0.1 %
Specific optical rotation -13.0° to -16.0° (20°C, 589 nm) (c=10, 2 N HCl)
Titration with HClO4 97.5 to 102.5 %
Other amino acids ≤0.5 % (TLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR