Inosine 5'-monophosphate disodium salt hydrate 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
5'-IMP 2Na hydrate, 5'-Inosinic acid disodium salt hydrate, Inosinic acid (disodium)(hydrate)(1:2:X), Inosine 5'-monophosphate disodium salt xhydrate, Disodium inosinate hydrate, Inosic acid disodium salt hydrate
22626-0050
22626-0250
22626-1000
TS22626-0050EA
37
USD
TS22626-0050
TS22626-0250
TS22626-1000
Inosine 5'-monophosphate disodium salt hydrate 97%
Inosine 5'-monophosphate disodium salt hydrate
Formula:
C₁₀H₁₁N₄Na₂O₈P·xH₂O MW: 392.17 g/mol (anhy) Melting Pt: 175 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00150372 CAS Number: 352195-40-5 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Water | ≤28.5 % (K.F.) |
UV spectrophotometry | molar absorptivity 12300 +/- 4%, on dry basis |
Heavy metals (as Pb) | ≤20 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...