Chromium 99+%, powder, pure

Supplier: Thermo Scientific Chemicals

225491000 225495000
TS225491000EA 53 USD
TS225491000 TS225495000
Chromium 99+%, powder, pure
Chromium
Formula: Cr
Boiling Pt: 2642 °C (760 mmHg)
Melting Pt: 1857 °C
Density: 7.2 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00010944
CAS Number: 7440-47-3

Order Now

Specification Test Results

Appearance Grey Metallic powder
Titration with Na2S2O3 ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR