Zinc sulfide 99.99% (trace metals basis) < 10 µm
Supplier: Thermo Scientific Chemicals
Synonyms:
Zinc (II) sulphide, Zinc (II) sulfide
AC22365-0250
AC22365-1000
AC22365-5000
TS22365-0250EA
39
USD
TS22365-0250
TS22365-1000
TS22365-5000
Zinc sulfide 99.99% (trace metals basis) < 10 µm
Zinc sulphide
Formula:
SZn MW: 97.46 g/mol Melting Pt: 1064 Storage Temperature: Ambient |
MDL Number:
MFCD00011301 CAS Number: 1314-98-3 EINECS: 215-251-3 Merck Index: 12,10294 |
Specification Test Results
Appearance | White to light yellow Powder |
Total trace metal impurities | ≤0.01 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...