tetra-Chloroauric(III) acid hydrate

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Gold(III) chloride hydrate, Hydrogen tetrachloroaurate(III) xhydrate, Hydrogen tetrachloroaurate (III) hydrate, Hydrogen tetrachloroaurate hydrate, Chloroauric acid, tetra-Chloroauric acid hydrate

AC22362-0010 AC22362-0050
TS22362-0010EA 231 USD
TS22362-0010 TS22362-0050
tetra-Chloroauric(III) acid hydrate
tetra-Chloroauric (III) acid hydrate
Formula: HAuCl₄·H₂O
Melting Pt: 30 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00149903
CAS Number: 27988-77-8
EINECS: 240-948-4
UN: 3260
ADR: 8,II

Order Now

Specification Test Results

Appearance Yellow to orange Powder or crystals and/or chunks
Gold (Au) ≥49 %
Insoluble matter ≤0.1 % (in diethyl ether)
Alkalies+oth.metals (SO4) ≤0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR