1-(Trimethylsilyl)-1-propyne 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Trimethyl-1-propynylsilane
AC22353-0010
AC22353-0050
TS22353-0010EA
36
USD
TS22353-0010
TS22353-0050
1-(Trimethylsilyl)-1-propyne 98%
1-(Trimethylsilyl)-1-propyne
Formula:
C₆H₁₂Si MW: 112.25 Boiling Pt: 99 Density: 0.75 g/cm³ Flash Pt: –3 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00009271 CAS Number: 6224-91-5 EINECS: 228-314-5 UN: 1993 ADR: 3 II |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4180 to 1.4200 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...