Sodium perborate tetrahydrate 97%

Supplier: Thermo Scientific Chemicals
Danger

AC22320-0010 AC22320-0050 AC22320-0250 AC22320-2500
TS22320-0010EA 129 USD
TS22320-0010 TS22320-0050 TS22320-0250 TS22320-2500
Sodium perborate tetrahydrate 97%
Sodium perborate tetrahydrate
Formula: NaH₂BO₄·4H₂O
Melting Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00149231
CAS Number: 13517-20-9
EINECS: 239-172-9

Order Now

Specification Test Results

Appearance White Crystalline powder
pH 10.1 to 10.6 (1 % solution)
Titration with KMnO4 ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR