N-Acetylethylenediamine 90%

Supplier: Thermo Scientific Chemicals
Danger

22317-0050
TS22317-0050EA 47 USD
TS22317-0050
N-Acetylethylenediamine 90%
N-Acetylethylenediamine
Formula: C₄H₁₀N₂O
Boiling Pt: 143 °C (10 mmHg)
Melting Pt: 50 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008163
CAS Number: 1001-53-2
UN: 3259
ADR: 8,III

Order Now

Specification Test Results

Appearance White to yellow Crystalline solid
Infrared spectrum Conforms
Melting point 45 to 50 °C
GC ≥88 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR