Tin(II) chloride dihydrate 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Stannous chloride dihydrate

AC22266-0025 AC22266-1000 AC22266-5000
TS22266-0025EA 645 USD
TS22266-0025 TS22266-1000 TS22266-5000
Tin(II) chloride dihydrate 98+%
Tin (II) chloride dihydrate
Formula: SnCl₂·2H₂O
Boiling Pt: 652 °C (760 mmHg)
Melting Pt: 37…38 °C
Storage Temperature: Ambient
MDL Number: MFCD00149863
CAS Number: 10025-69-1
EINECS: 231-868-0
UN: 3260
ADR: 8,III
Merck Index: 13,08861

Order Now

Specification Test Results

Appearance Colorless to white Crystalline powder or crystals and/or chunks
Titration Iodimetric 98.0 to 103.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR