Zirconyl nitrate hydrate 99.5% contains up to 4% Hf
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Zirconium(IV) oxynitrate hydrate, Zirconium(IV) dinitrate oxide hydrate, Zirconium dinitrate oxide hydrate
AC22263-1000
AC22263-5000
TS22263-1000EA
104
USD
TS22263-1000
TS22263-5000
Zirconyl nitrate hydrate 99.5% contains up to 4% Hf
Zirconyl nitrate hydrate
Formula:
N₂O₇Zr·H₂O |
MDL Number:
MFCD00149896 CAS Number: 14985-18-3 EINECS: 237-529-3 UN: 3085 ADR: 5.1,III |
Specification Test Results
Appearance | White Powder and/or chunks |
Degree of hydration | 6 to 7 |
Gravimetry | 34.9 to 37.0 % (ZrO2) (calculated on the actual degree of hydration) |
Hafnium (Hf) | ≤4 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...