Zinc, granular 20 mesh

Supplier: Thermo Scientific Chemicals
Danger

222605000 222600025
TS222605000EA 71 USD
TS222605000 TS222600025
Zinc, granular 20 mesh
Zinc
Formula: Zn
MW: 65.39 g/mol
Boiling Pt: 908
Melting Pt: 420
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
UN: 1436
Merck Index: 12,10255

Order Now

Specification Test Results

Appearance Silver grey Granules or filings
Titration Complexometric ≥99.8 %
Particle size 20 mesh
Suitability (For determination of As) Passes test
Iron (Fe) ≤0.01 %
Lead (Pb) ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR