Poly(styrene sulfonic acid) sodium salt, M.W. ∼ 70.000

Supplier: Thermo Scientific Chemicals

22227-0050 222271000 222275000
TS22227-0050EA 36 USD
TS22227-0050 TS222271000 TS222275000
Poly(styrene sulfonic acid) sodium salt, M.W. ∼ 70.000
Poly(styrene sulfonic acid) sodium salt

Appearance: White to tan Powder

Formula: [-CH₂CH(C₆H₄)SO₃Na)-]n
Storage Temperature: Ambient
MDL Number: MFCD00084449
CAS Number: 25704-18-1

Order Now

Specification Test Results

Appearance White to tan Powder
Infrared spectrum Conforms


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR