4-Bromo-N,N-dimethylaniline 99%

Supplier: Thermo Scientific Chemicals
Warning

22221-0250 22221-1000
TS22221-0250EA 43 USD
TS22221-0250 TS22221-1000
4-Bromo-N,N-dimethylaniline 99%
4-Bromo-N,N-dimethylaniline
Formula: C₈H₁₀BrN
Boiling Pt: 264 °C (1013 hPa)
Melting Pt: 53…56 °C
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00000093
CAS Number: 586-77-6
EINECS: 209-582-2
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to grey to light blue Crystalline solid
Infrared spectrum Conforms
Melting point 53 to 56 °C
Titration with HClO4 ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR