Tetrahydrofuran 99.5+% for spectroscopy

Supplier: Thermo Scientific Chemicals
Danger

22216-2500 22216-5000 222160010 222160025
TS22216-2500EA 72 USD
TS22216-2500 TS22216-5000 TS222160010 TS222160025
Tetrahydrofuran 99.5+% for spectroscopy
Tetrahydrofuran
Formula: C₄H₈O
Boiling Pt: 66 °C (760 mmHg)
Melting Pt: –108.4 °C
Density: 0.88 g/cm³
Flash Pt: –21 °C
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3,II
REACH: 05-2114615188-48
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.03 % (Coulometric)
GC ≥99.50 %
UV 1 cm cell vs HPLC-grade water
Color scale ≤10 APHA
Residue after evaporation ≤0.005 %
Acidity ≤0.0002 meq/g
Alkalinity ≤0.0006 meq/g
Peroxides ≤0.01 % (as H2O2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR