Diphenylamine 99+%, extra pure indicator grade, redox indicator

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N-Phenylaniline, N-Phenylbenzeneamine

22192-1000 22192-5000
TS22192-1000EA 75 USD
TS22192-1000 TS22192-5000
Diphenylamine 99+%, extra pure indicator grade, redox indicator
Diphenylamine
Formula: (C₆H₅)₂NH
MW: 169.23 g/mol
Boiling Pt: 302
Melting Pt: 52…54 °C
Flash Pt: 152 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003014
CAS Number: 122-39-4
EINECS: 204-539-4
UN: 3077
ADR: 9 III
Merck Index: 13,03349
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23b05bdc0d251be7093ced24f3f9389bc8e8d3c5&vid=f543cfadbe22d37b18bd37917bb8b5672a6d2df4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals or flakes
Infrared spectrum Conforms
Sulfated ash ≤0.05 %
GC ≥99.0 %
Sensitivity (to nitrate) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR