Copper(II) oxide 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Cupric oxide, Copper oxide, Copper monoxide granulated

AC22128-0025 AC22128-1000 AC22128-5000
TS22128-0025EA 903 USD
TS22128-0025 TS22128-1000 TS22128-5000
Copper(II) oxide 99+%
Copper (II) oxide
Formula: CuO
MW: 79.55 g/mol
Boiling Pt: 2000
Melting Pt: 1326
Storage Temperature: Ambient
MDL Number: MFCD00010979
CAS Number: 1317-38-0
EINECS: 215-269-1
UN: 3077
ADR: 9 III
Merck Index: 13,02674

Order Now

Specification Test Results

Appearance Black Powder
Titration Complexometric ≥99.0 %
Chloride (Cl) ≤50 ppm
Carbon (C) ≤500 ppm
Iron (Fe) ≤500 ppm
Total sulfur (as SO4) ≤0.02 %
Free alkali Passes test
Insoluble matter ≤0.02 % (in dilute HCl)
Nitrogen compounds (as N) ≤0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR