Hydroquinone 99.5%
Supplier: Thermo Scientific Chemicals
|
Danger
|
21993-0051
21993-0010
21993-0500
21993-2500
TS21993-0051EA
349
USD
TS21993-0051
TS21993-0010
TS21993-0050
TS21993-0500
TS21993-2500
Hydroquinone 99.5%
Hydroquinone
Formula:
C₆H₄(OH)₂ Boiling Pt: 285…287 °C (760 mmHg) Melting Pt: 170…174 °C Flash Pt: 165 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002339 CAS Number: 123-31-9 EINECS: 204-617-8 UN: 3077 ADR: 9,III Merck Index: 13,04833 |
|
Specification Test Results
Appearance | White to off-white Crystalline powder or crystals or needles |
Infrared spectrum | Conforms |
Melting point | 171 to 174 °C |
Water | ≤0.6 % (K.F.) |
Sulfated ash | ≤0.05 % |
HPLC | ≥99.4 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...