Hydroquinone 99.5%

Supplier: Thermo Scientific Chemicals
Danger

21993-0051 21993-0010 21993-0500 21993-2500
TS21993-0051EA 349 USD
TS21993-0051 TS21993-0010 TS21993-0050 TS21993-0500 TS21993-2500
Hydroquinone 99.5%
Hydroquinone
Formula: C₆H₄(OH)₂
Boiling Pt: 285…287 °C (760 mmHg)
Melting Pt: 170…174 °C
Flash Pt: 165 °C
Storage Temperature: Ambient
MDL Number: MFCD00002339
CAS Number: 123-31-9
EINECS: 204-617-8
UN: 3077
ADR: 9,III
Merck Index: 13,04833
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=203cfb319003b2b4489cbe3ab15876fc37f27cb8&vid=6d76260c677491e292c03637f8a08e9194282d48&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals or needles
Infrared spectrum Conforms
Melting point 171 to 174 °C
Water ≤0.6 % (K.F.)
Sulfated ash ≤0.05 %
HPLC ≥99.4 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR