Benzophenone 99+%, pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Diphenylmethanone, Diphenyl ketone

21968-0010 21968-0050 21968-0500 21968-2500
TS21968-0010EA 98 USD
TS21968-0010 TS21968-0050 TS21968-0500 TS21968-2500
Benzophenone 99+%, pure
Benzophenone
Formula: C₁₃H₁₀O
Boiling Pt: 305.4 °C (1013 hPa)
Melting Pt: 47…49 °C
Flash Pt: 143 °C
Storage Temperature: Ambient
MDL Number: MFCD00003076
CAS Number: 119-61-9
EINECS: 204-337-6
Merck Index: 13,01099

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or flakes
Infrared spectrum Conforms
Melting point 47 to 49 °C
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR