Acetyl chloride 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Acetyl chloride 1M in Dichloromethane

21947-0250 21947-2500 21947-0010 21947-0025
TS21947-0250EA 61 USD
TS21947-0250 TS21947-2500 TS21947-0010 TS21947-0025
Acetyl chloride 99+%
Acetyl chloride
Formula: C₂H₃ClO
Boiling Pt: 51 °C (760 mmHg)
Melting Pt: –112 °C
Density: 1.1 g/cm³
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00000719
CAS Number: 75-36-5
EINECS: 200-865-6
UN: 1717
ADR: 3,II
Merck Index: 13,00087

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3875 to 1.3895 (20°C, 589 nm)
GC ≥99.0 %
Color scale ≤20 APHA
Titration RCOCl 99.0 to 101.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR