Acetyl chloride 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Acetyl chloride 1M in Dichloromethane
21947-0250
21947-2500
21947-0010
21947-0025
TS21947-0250EA
61
USD
TS21947-0250
TS21947-2500
TS21947-0010
TS21947-0025
Acetyl chloride 99+%
Acetyl chloride
Formula:
C₂H₃ClO Boiling Pt: 51 °C (760 mmHg) Melting Pt: –112 °C Density: 1.1 g/cm³ Flash Pt: 4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000719 CAS Number: 75-36-5 EINECS: 200-865-6 UN: 1717 ADR: 3,II Merck Index: 13,00087 |
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3875 to 1.3895 (20°C, 589 nm) |
GC | ≥99.0 % |
Color scale | ≤20 APHA |
Titration RCOCl | 99.0 to 101.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...